Skip to Content
Merck
SAHA ≥98% (HPLC)

SAHA

SAHA ≥98% (HPLC)
Synonym(s):
N-hydroxy-N′-phenyl-octanediamide, Suberoylanilide hydroxamic acid, Vorinostat
Empirical Formula (Hill Notation):
C14H20N2O3
CAS Number:
Molecular Weight:
264.32

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing