Skip to Content
Merck
(S)-2-Phenyl-1-propanol purum, ≥98.0% (sum of enantiomers, GC)

(S)-2-Phenyl-1-propanol

(S)-2-Phenyl-1-propanol purum, ≥98.0% (sum of enantiomers, GC)
Synonym(s):
(S)-β-Methylphenethyl alcohol
Linear Formula:
CH3CH(C6H5)CH2OH
CAS Number:
Molecular Weight:
136.19

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing