Skip to Content
Merck
S-(2-Hydroxy-5-nitrobenzyl)-6-thioinosine 97%

S-(2-Hydroxy-5-nitrobenzyl)-6-thioinosine

S-(2-Hydroxy-5-nitrobenzyl)-6-thioinosine 97%
Synonym(s):
6-[(2-Hydroxy-5-nitrobenzyl)thio]-9-β-D-ribofuranosylpurine
Empirical Formula (Hill Notation):
C17H17N5O7S
CAS Number:
Molecular Weight:
435.41

Products

Brand
Formula Weight
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing