Residual Solvent Class 2 - 1,4-Dioxane
Synonym(s):
1,4-Dioxane solution
Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing