![Ranolazine Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material, certified reference material, pharmaceutical secondary standard, pkg of 100 mg](/deepweb/assets/sigmaaldrich/product/images/320/774/b13e314e-caf0-4a0d-a791-66e3f77a92b5/640/b13e314e-caf0-4a0d-a791-66e3f77a92b5.jpg)
Ranolazine Related Compound A
![Ranolazine Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material, certified reference material, pharmaceutical secondary standard, pkg of 100 mg](/deepweb/assets/sigmaaldrich/product/images/320/774/b13e314e-caf0-4a0d-a791-66e3f77a92b5/640/b13e314e-caf0-4a0d-a791-66e3f77a92b5.jpg)
Synonym(s):
(±)-2-[(2-Methoxyphenoxy)methyl]oxirane, NSC 112256, NSC 133442
Empirical Formula (Hill Notation):
C10H12O3
CAS Number:
Molecular Weight:
180.20
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing