
Ranitidine Impurity K

Synonym(s):
N-Methyl-1-(methylthio)-2-nitroethenamine
Linear Formula:
O2NCH=C(SCH3)NHCH3
CAS Number:
Molecular Weight:
148.18
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing