Skip to Content
Merck
Ractopamine Hydrochloride certified reference material, pharmaceutical secondary standard

Ractopamine hydrochloride

Ractopamine Hydrochloride certified reference material, pharmaceutical secondary standard
Synonym(s):
4-{3-[2-Hydroxy-2-(4-hydroxyphenyl)ethyl]aminobutylphenol hydrochloride, Ractopamine hydrochloride
Empirical Formula (Hill Notation):
C18H23NO3 · HCl
CAS Number:
Molecular Weight:
337.84

Products

Brand
Formula Weight
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique