Skip to Content
Merck
Quinine sulfate meets USP testing specifications, monograph mol wt. 782.94 ((C20H24N2O2)2 . H2SO4 . 2H2O)

Quinine sulfate

Quinine sulfate meets USP testing specifications, monograph mol wt. 782.94 ((C20H24N2O2)2 . H2SO4 . 2H2O)
Synonym(s):
Quinine hemisulfate salt monohydrate, Quinine sulfate (2:1) (salt) dihydrate
Empirical Formula (Hill Notation):
C20H24N2O2 · 0.5H2O4S · H2O
CAS Number:
Molecular Weight:
391.47

Products

Feature
Formula Weight
Manufacturer
Melting Point (°C)
Activity Spectrum
Agency
Format
Application
Markush Class
Markush Group
Mode of Action
API Family
Special Grade