Skip to Content
Merck
Pyriftalid PESTANAL®, analytical standard

Pyriftalid

Pyriftalid PESTANAL®, analytical standard
Synonym(s):
7-(4,6-Dimethoxypyrimidin-2-ylthio)-3-methylisobenzofuran-1(3H)-one
Empirical Formula (Hill Notation):
C15H14N2O4S
CAS Number:
Molecular Weight:
318.35

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing