Prochloraz Metabolite BTS40348
Synonym(s):
N-[2-(2,4,6-Trichlorophenoxy)ethyl]propylamine, N-Propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]amine
Empirical Formula (Hill Notation):
C11H14Cl3NO
CAS Number:
Molecular Weight:
282.59
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing