![Poly[(phenyl isocyanate)-co-formaldehyde] average Mn ~400](/deepweb/assets/sigmaaldrich/product/structures/319/634/60a69bae-e14c-4356-983b-a2c362dcdeec/640/60a69bae-e14c-4356-983b-a2c362dcdeec.png)
Poly[(phenyl isocyanate)-co-formaldehyde]
![Poly[(phenyl isocyanate)-co-formaldehyde] average Mn ~400](/deepweb/assets/sigmaaldrich/product/structures/319/634/60a69bae-e14c-4356-983b-a2c362dcdeec/640/60a69bae-e14c-4356-983b-a2c362dcdeec.png)
Synonym(s):
PMDI, Polymethylene polyphenyl isocyanate
Linear Formula:
[C6H3(NCO)CH2]n
CAS Number:
Products
Brand
Boiling Point (°C)
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing