Poly(N-isopropylacrylamide-co-methacrylic acid)
Synonym(s):
Poly(NIPAM-co-MAA), Polyacrylamide, functionalized polyNIPAM, functionalized polyacrylamide, polyNIPAM
Linear Formula:
(C6H11NO)m (C4H6O2)n
CAS Number:
Products
Brand
Melting Point (°C)
Physical Form
Chemical Composition
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing