![Piperophos PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/348/817/7d51e716-365a-4501-b8d1-cf51a132c2b6/640/7d51e716-365a-4501-b8d1-cf51a132c2b6.png)
Piperophos
![Piperophos PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/348/817/7d51e716-365a-4501-b8d1-cf51a132c2b6/640/7d51e716-365a-4501-b8d1-cf51a132c2b6.png)
Empirical Formula (Hill Notation):
C14H28NO3PS2
CAS Number:
Molecular Weight:
353.48
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing