Phenylpropanolamine hydrochloride
Synonym(s):
DL-Norephedrine hydrochloride, (±)-Phenylpropanolamine hydrochloride, erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-Phenylpropanolamine hydrochloride
Linear Formula:
C6H5CH(OH)CH(NH2)CH3 · HCl
CAS Number:
Molecular Weight:
187.67
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing