
Palmitic acid-2,2-d2

Synonym(s):
Hexadecanoic-2,2-d2 acid
Linear Formula:
CH3(CH2)13CD2CO2H
CAS Number:
Molecular Weight:
258.44
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing