![Oxymatrine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/411/332/8a973b81-4369-499b-8354-d128cf6c238a/640/8a973b81-4369-499b-8354-d128cf6c238a.png)
Oxymatrine
![Oxymatrine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/411/332/8a973b81-4369-499b-8354-d128cf6c238a/640/8a973b81-4369-499b-8354-d128cf6c238a.png)
Empirical Formula (Hill Notation):
C15H24N2O2
CAS Number:
Molecular Weight:
264.36
Products
Formula Weight
Manufacturer
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing