Skip to Content
Merck
(Oxydi-2,1-phenylene)bis(diphenylphosphine) 98%

(Oxydi-2,1-phenylene)bis(diphenylphosphine)

(Oxydi-2,1-phenylene)bis(diphenylphosphine) 98%
Synonym(s):
Bis[(2-diphenylphosphino)phenyl] ether, DPEPhos
Linear Formula:
O[C6H4P(C6H5)2]2
CAS Number:
Molecular Weight:
538.55

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing