Skip to Content
Merck
o-Toluenethiol ≥97%, FG

o-Toluenethiol

o-Toluenethiol ≥97%, FG
Synonym(s):
2-Methylbenzenethiol, o-Thiocresol
Linear Formula:
CH3C6H4SH
CAS Number:
Molecular Weight:
124.20
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing