![o-Phenylene phosphorochloridite 97%](/deepweb/assets/sigmaaldrich/product/structures/238/590/fce98c80-6318-4cd6-91e6-1e9497258330/640/fce98c80-6318-4cd6-91e6-1e9497258330.png)
o-Phenylene phosphorochloridite
![o-Phenylene phosphorochloridite 97%](/deepweb/assets/sigmaaldrich/product/structures/238/590/fce98c80-6318-4cd6-91e6-1e9497258330/640/fce98c80-6318-4cd6-91e6-1e9497258330.png)
Synonym(s):
2-Chloro-1,3,2-benzodioxaphosphole, o-Phenylene chlorophosphite, Chloro(1,2-phenylenedioxy)phosphine, Cyclic o-phenylene phosphorochloridite, Phosphorochloridous acid orthophenylene ester, Pyrocatechol phosphoryl chloride
Empirical Formula (Hill Notation):
C6H4ClO2P
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing