![n-Propylmagnesium chloride solution 1.0 M in 2-methyltetrahydrofuran](/deepweb/assets/sigmaaldrich/product/structures/791/902/9503b177-aac5-43f3-9bb8-aa5b64d89b3b/640/9503b177-aac5-43f3-9bb8-aa5b64d89b3b.png)
n-Propylmagnesium chloride solution
![n-Propylmagnesium chloride solution 1.0 M in 2-methyltetrahydrofuran](/deepweb/assets/sigmaaldrich/product/structures/791/902/9503b177-aac5-43f3-9bb8-aa5b64d89b3b/640/9503b177-aac5-43f3-9bb8-aa5b64d89b3b.png)
Synonym(s):
Propylmagnesium chloride solution
Linear Formula:
CH3CH2CH2MgCl
CAS Number:
Molecular Weight:
102.85
Products
Brand
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing