![N,N-Dimethyltryptamine ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/236/418/3425ab09-0209-41be-a482-0206d9d054bd/640/3425ab09-0209-41be-a482-0206d9d054bd.png)
N,N-Dimethyltryptamine
![N,N-Dimethyltryptamine ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/236/418/3425ab09-0209-41be-a482-0206d9d054bd/640/3425ab09-0209-41be-a482-0206d9d054bd.png)
Synonym(s):
DMT, Dimethyltryptamine, N,N-Dimethyl-1H-indole-3-ethanamine
Empirical Formula (Hill Notation):
C12H16N2
CAS Number:
Molecular Weight:
188.27
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing