![Nikethamide European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/318/135/48613f21-f289-4a46-9bd0-74b2f11e6519/640/48613f21-f289-4a46-9bd0-74b2f11e6519.png)
Nikethamide
![Nikethamide European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/318/135/48613f21-f289-4a46-9bd0-74b2f11e6519/640/48613f21-f289-4a46-9bd0-74b2f11e6519.png)
Synonym(s):
N,N-Diethylnicotinamide, Nicotinic acid diethylamide, ‘Nikethamide’
Empirical Formula (Hill Notation):
C10H14N2O
CAS Number:
Molecular Weight:
178.23
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing