![N-Debenzoyl-N-(phenylacetyl)paclitaxel certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/363/515/47a10928-a69f-4793-9179-ca5677e80447/640/47a10928-a69f-4793-9179-ca5677e80447.png)
N-Debenzoyl-N-(phenylacetyl)paclitaxel
![N-Debenzoyl-N-(phenylacetyl)paclitaxel certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/363/515/47a10928-a69f-4793-9179-ca5677e80447/640/47a10928-a69f-4793-9179-ca5677e80447.png)
Empirical Formula (Hill Notation):
C48H53NO14
CAS Number:
Molecular Weight:
867.93
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing