Skip to Content
Merck
N-Boc-hydroxylamine ≥98%

N-Boc-hydroxylamine

N-Boc-hydroxylamine ≥98%
Synonym(s):
tert-Butyl N-hydroxycarbamate
Linear Formula:
(CH3)3COCONHOH
CAS Number:
Molecular Weight:
133.15
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing