Skip to Content
Merck
vibrant-m

Nabumetone impurity D

vibrant-m
Synonym(s):
(E)-4-(6-Methoxynaphthalen-2-yl)but-3-en-2-one
Empirical Formula (Hill Notation):
C15H14O2
CAS Number:
Molecular Weight:
226.27

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade