Skip to Content
Merck
N-[(1R,3r,5S)-9-Azabicyclo[3.3.1]non-3-yl]-1-methyl-1H-indazole-3-carboxamide monohydrochloride certified reference material, pharmaceutical secondary standard

N-[(1R,3r,5S)-9-Azabicyclo[3.3.1]non-3-yl]-1-methyl-1H-indazole-3-carboxamide monohydrochloride

N-[(1R,3r,5S)-9-Azabicyclo[3.3.1]non-3-yl]-1-methyl-1H-indazole-3-carboxamide monohydrochloride certified reference material, pharmaceutical secondary standard
Synonym(s):
N-Demethyl-endo-granisetron hydrochloride, endo-N-9-Azabicyclo[3.3.1]non-3-yl-1-methyl-1H-indazole-3-carboxamide monohydrochloride
Empirical Formula (Hill Notation):
C17H22N4O · HCl
CAS Number:
Molecular Weight:
334.84

Products

Brand
Formula Weight
Quality Segment
Application
Package Size
API Family
Special Grade