Skip to Content
Merck
Myelin Basic Protein Guinea Pig Fragment 68-82 ≥97% (HPLC)

Myelin Basic Protein Guinea Pig Fragment 68-82

Myelin Basic Protein Guinea Pig Fragment 68-82 ≥97% (HPLC)
Synonym(s):
des-Gly77, des-His78-Myelin Basic Protein Bovine fragment 68-84
Empirical Formula (Hill Notation):
C71H113N23O28
CAS Number:
Molecular Weight:
1736.79

Products

Product Category
Brand
Formula Weight
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing