
Monoclonal Anti-CDC25C antibody produced in mouse

Synonym(s):
Anti-CDC25, Anti-cell division cycle 25C
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2