Skip to Content
Merck
Mogrol phyproof® Reference Substance

Mogrol

Mogrol phyproof® Reference Substance
Synonym(s):
Mogrol, (3β,9β,10α,11α,24R)-9-Methyl-19-norlanost-5-ene-3,11,24,25-tetrol
Empirical Formula (Hill Notation):
C30H52O4
CAS Number:
Molecular Weight:
476.73

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing