Skip to Content
Merck
Methylene Blue Chloride Harleco®, suitable for histology

Methylene Blue Chloride

Methylene Blue Chloride Harleco®, suitable for histology
Synonym(s):
Methylthioninium chloride, Methylene blue
Empirical Formula (Hill Notation):
C16H18ClN3S
CAS Number:
Molecular Weight:
319.85

Products

Brand
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing