Skip to Content
Merck
Methyl 4-hydroxybenzoate analytical standard

Methyl 4-hydroxybenzoate

Methyl 4-hydroxybenzoate analytical standard
Synonym(s):
p-Hydroxybenzoic acid methyl ester, Methyl paraben, NIPAGIN, Methyl 4-hydroxybenzoate
Linear Formula:
HOC6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
EC Number:

Products

Product Category
Brand
Biological Source
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Research Area
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Package Size
Special Grade
Technique