Skip to Content
Merck
Methyl 2-amino-4,5-dimethoxybenzoate ≥98.0% (HPLC/NT)

Methyl 2-amino-4,5-dimethoxybenzoate

Methyl 2-amino-4,5-dimethoxybenzoate ≥98.0% (HPLC/NT)
Synonym(s):
Methyl 4,5-dimethoxyanthranilate, Methyl 6-aminoveratrate
Empirical Formula (Hill Notation):
C10H13NO4
CAS Number:
Molecular Weight:
211.21
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing