![MES potassium salt ≥99% (titration)](/deepweb/assets/sigmaaldrich/product/structures/161/323/2ab392ca-be3b-426a-855f-e09ee1826204/640/2ab392ca-be3b-426a-855f-e09ee1826204.png)
MES potassium salt
![MES potassium salt ≥99% (titration)](/deepweb/assets/sigmaaldrich/product/structures/161/323/2ab392ca-be3b-426a-855f-e09ee1826204/640/2ab392ca-be3b-426a-855f-e09ee1826204.png)
Synonym(s):
2-(N-Morpholino)ethanesulfonic acid potassium salt, 4-Morpholineethanesulfonic acid potassium salt
Empirical Formula (Hill Notation):
C6H12KNO4S
CAS Number:
Molecular Weight:
233.33
Products
Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
pK Value
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing