
Mercury(II) nitrate monohydrate

Synonym(s):
Mercury(II) nitrate monohydrate
Linear Formula:
Hg(NO3)2 · H2O
CAS Number:
Molecular Weight:
324.60 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing