Meperidine hydrochloride
Synonym(s):
1-Methyl-4-phenylpiperidine-4-carboxylic acid ethyl ester hydrochloride, Pethidine hydrochloride
Empirical Formula (Hill Notation):
C15H21NO2 · HCl
CAS Number:
Molecular Weight:
283.79
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3