Skip to Content
Merck
Mandipropamid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Mandipropamid

Mandipropamid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-(4-Chlorophenyl)-N-[3-methoxy-4-(2-propynyloxy)phenethyl]-2-(2-propynyloxy)acetamide, (±)-Chlorophenyl)-N-[2-[3-methoxy-4-(prop-2-ynyloxy)phenyl]ethyl]-2-(prop-2-ynyloxy)acetamide, 4-Chloro-N-[2-[3-methoxy-4-(2-propyn-1-yloxy)phenyl]ethyl]-α-(2-propyn-1-yloxy)benzeneacetamide, Mandi
Empirical Formula (Hill Notation):
C23H22ClNO4
CAS Number:
Molecular Weight:
411.88

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique