Magnesium chloride
Synonym(s):
Magnesium chloride hexahydrate
Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2