Skip to Content
Merck
MacConkey Agar with Crystal violet, Sodium chloride and 0.15% Bile salts NutriSelect® Plus, suitable for microbiology

MacConkey Agar with Crystal violet, Sodium chloride and 0.15% Bile salts

MacConkey Agar with Crystal violet, Sodium chloride and 0.15% Bile salts NutriSelect® Plus, suitable for microbiology

Products

Shipping
Product Category
pH Value
Physical Form
Quality Segment
Application
Microorganism Suitability
Sterility
Technique