Loperamide N-oxide
Synonym(s):
trans-4-(4-Chlorophenyl)-4-hydroxy-N,N-dimethyl-α,α-diphenyl-1-piperidinebutanamide 1-oxide
Empirical Formula (Hill Notation):
C29H33ClN2O3
CAS Number:
Molecular Weight:
493.04
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing