Skip to Content
Merck
L-Lysine ≥98%, FG

L-Lysine

L-Lysine ≥98%, FG
Synonym(s):
(S)-2,6-Diaminocaproic acid
Linear Formula:
H2N(CH2)4CH(NH2)CO2H
CAS Number:
Molecular Weight:
146.19
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Analyte Suitability
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4