![Limonin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/362/910/7317a617-e61b-4084-bdbf-ec26dfe27cce/640/7317a617-e61b-4084-bdbf-ec26dfe27cce.png)
Limonin
![Limonin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/362/910/7317a617-e61b-4084-bdbf-ec26dfe27cce/640/7317a617-e61b-4084-bdbf-ec26dfe27cce.png)
Synonym(s):
Citrolimonin, Dictamnolactone, Evodin, Limonoic acid 3,19:16,17 dilactone, Obaculactone
Empirical Formula (Hill Notation):
C26H30O8
CAS Number:
Molecular Weight:
470.51
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Tissue
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing