
Levodropropizine impurity C

Synonym(s):
(±)-Glycidol, (±)-Oxirane-2-methanol, 2,3-Epoxy-1-propanol, Glycerolglycide
Empirical Formula (Hill Notation):
C3H6O2
CAS Number:
Molecular Weight:
74.08
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing