Skip to Content
Merck
Lacosamide ≥98% (HPLC)

Lacosamide

Lacosamide ≥98% (HPLC)
Synonym(s):
(2R)-2-(Acetylamino)-N-benzyl-3-methoxypropanamide, ADD 243037, ADD-243037, ADD243037, Erlosamide, LCM, SPM-927, SPM927
Empirical Formula (Hill Notation):
C13H18N2O3
CAS Number:
Molecular Weight:
250.29

Products

Brand
Color
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing