Skip to Content
Merck
Isoprene analytical standard

Isoprene

Isoprene analytical standard
Synonym(s):
2-Methyl-1,3-butadiene
Linear Formula:
CH2=CHC(CH3)=CH2
CAS Number:
Molecular Weight:
68.12
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing