Skip to Content
Merck
Isethionic acid ammonium salt 99%

Isethionic acid ammonium salt

Isethionic acid ammonium salt 99%
Synonym(s):
2-Hydroxyethanesulfonic acid ammonium salt
Linear Formula:
HOCH2CH2SO3NH4
CAS Number:
Molecular Weight:
143.16
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing