Skip to Content
Merck
Indeno[1,2-b]fluorene-6,12-dione 99%

Indeno[1,2-b]fluorene-6,12-dione

Indeno[1,2-b]fluorene-6,12-dione 99%
Synonym(s):
trans-Fluorenacenedione, Indenofluorenedione
Empirical Formula (Hill Notation):
C20H10O2
CAS Number:
Molecular Weight:
282.29

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing