![Hydrocortisone Impurity L Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/115/025/962a846b-af1b-48f5-a716-c3479e71598a/640/962a846b-af1b-48f5-a716-c3479e71598a.png)
Hydrocortisone Impurity L
![Hydrocortisone Impurity L Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/115/025/962a846b-af1b-48f5-a716-c3479e71598a/640/962a846b-af1b-48f5-a716-c3479e71598a.png)
Synonym(s):
21-Deoxycortisol, 11β,17α-Dihydroxy-4-pregnene-3,20-dione, 11β,17α-Dihydroxyprogesterone, 21-Desoxycortisol, 4-Pregnene-11β,17α-diol-3,20-dione
Empirical Formula (Hill Notation):
C21H30O4
CAS Number:
Molecular Weight:
346.46
Products
Brand
Formula Weight
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing