
HMMNI-d3

Synonym(s):
2-Hydroxymethyl-1-methyl-d3-5-nitro-1H-imidazole
Empirical Formula (Hill Notation):
C5H4D3N3O3
CAS Number:
Molecular Weight:
160.15
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing