Guanidinium chloride
Synonym(s):
Guanidine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidinium chloride
Linear Formula:
NH2C(=NH)NH2 · HCl
CAS Number:
Molecular Weight:
95.53
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Product Line
Agency
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing