Grindelic acid
Empirical Formula (Hill Notation):
C20H32O3
CAS Number:
Molecular Weight:
320.47
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing